Learn More
Pluriselect pluriStrainer (Cell Strainer) 40 um mesh-size PET, single packed, sterile, 25/pk
Supplier: Pluriselect 435004003
The Flip-Strainer Brand “Pluristrainer” is a separation device (or cell strainer) for a wide range of laboratory applications where the filtration, particle separation and the purification of liquids is required. Its unique design features improved ventilation to avoid clogging for a smart sample preparation. Pluristrainers are stackable to a cascade of strainers. They are flip-strainers and allow flashing back the separated particles. For large sample volumes it can be combined with a funnel. It fits all major brand 50 mL centrifuge . (conical) tubes. The combination with the Connector Ring allows the evacuation of the 50 ml tube-Strainer unit to increase the flow rate. Pressure exchange can be completely blocked. The sample will now stay inside the strainer and can be incubated/treated, including with commonly used cell lysis reagents. This product line cannot be autoclaved!
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.