Learn More
GeneCopoeia PLP ATXN2 (NM_002973.4
SDP
Supplier:  GeneCopoeia LPPX1075LV120200
Encompass
purified lentiviral particles for ATXN2 (NM_002973.4) codon optimized version with CMV promoter, C-3xHA tag, puromycin (50uL x 4 vials, >10^8 TUs/mL)
Catalog No. NC1753000