Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Verax Biomedical™ Platelet PGD™ Test System

Rapid test for the detection of Gram-positive and Gram-negative bacterial contamination in platelets. FDA cleared as a Safety Measure allowing the extension of platelet dating from 5 to 7 days when testing leukocyte reduced apheresis platelets suspended in plasma.

Specifications

Content And Storage 15°C to 30°C and 2°C to 8°C
CE Marker No
Clia Complexity Moderate
Format Lateral Flow
Sample Type Platelets
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
23-051-000
View Documents
Verax Biomedical™
P100/DISCBYVEND
100 Tests This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
23-051-001
View Documents
Verax Biomedical™
P20/DISCBYVEND
20 Tests This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
Description

Description

  • Visual, three-step test for the qualitative detection of bacterial contamination
  • Detects aerobes and anaerobes
  • Detection rate: 1:3069 in early culture screened leukoreduced apheresis platelets
  • Specificity: 99.9% in leukoreduced apheresis platelets
  • 500uL sample size
  • Includes disposables required to perform the test
  • Ready to use liquid reagents in dropper bottles
  • Gram-positive and Gram-negative read windows
  • Colored line in read window indicates a reactive result
  • Built in procedural controls verify assay validity
  • FDA cleared for use with: leukoreduced apheresis platelets in plasma or PAS-C, pools of up to 6 pre-storage or post storage pooled platelets and individual WBD platelets
  • CLIA Complexity: Moderate
Specifications

Specifications

15°C to 30°C and 2°C to 8°C
Moderate
Platelets
500 μL
No
Lateral Flow
Carton
Safety and Handling

Safety and Handling

ShelfLife : >3 Months

SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit