Learn More
Chemglass Life Sciences Plate glass, disc, borosilicate, 4in Diameter X 1/4in Thick
Supplier: Chemglass Life Sciences CG190617
Plate glass, disc, borosilicate, 4in Diameter X 1/4in Thick. Pyrex or Duran borosilicate glass disc. Edges on all sizes are ground and chamfered.