Learn More
Uline Plastic Drum Dolly - 55 Gallon,1 EA
Supplier: Uline H9757/3+
Move 55-gallon drums up to 900 lbs. with ease. Resists rust, denting. Solid bottom contains spills. 2" lip frames help stabilize drums. Five 3 x 1 " polyolefin swivel casters provide greater mobility.