Learn More
EUROTROL Plasma Low Hb Level 2
Supplier: EUROTROL 139.002.002
This specially formulated matrix mimics the characteristics of fresh human plasma and offers: Three physiological levels. A long shelf life (at 2 - 8°C) to deliver test consistency. 30-day open vial stability at room temperature, helping to ensure QC compliance.
This item is not returnable.
View return policy