Learn More
Darwin Chambers Company LED growth light fixture. Uniform light 2' x 2' tray fixture
SDP
Supplier:  Darwin Chambers Company PGLED
Encompass_Preferred
LED growth light fixture. Uniform light 2' x 2' tray fixture. 4000k temperature, approx. 1000 µmoles/m²/s manually dimming capable. Light timer sold separately. Separate 15amp 120V dedicated circuit may be required.
Catalog No. NC1635405