Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ Seamless Knit Cotton/Polyester Gloves with Double-Sided PVC Dense Dot Grip

Catalog Number 191300861
Click to view available options
Large
This item is not returnable. View return policy
This item is not returnable. View return policy

PVC dots on both sides of the glove enhance grip capabilities, as well as extend the useful life of the gloves. Knit wrist helps prevent dirt and debris from entering the glove.

  • Seamless knit shell is comfortable and flexible
  • Knit wrist helps prevent dirt and debris from entering the glove
  • Use in warehousing, package handling, general assembly, light duty general work situations, and construction
  • Size small is ideal fit for women
Construction Seamless Knit
Coating Material PVC Dots (2 Sides)
Color Natural, Blue
Hand Ambidextrous
Material Polycotton
Product Type Seamless Knit Gloves
Size Large
For Use With (Application) Warehousing, Package Handling, General Assembly, and Construction
Glove Cuff Style Knit
Series 37-C512PDD

WARNING: Cancer - www.P65Warnings.ca.gov

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit