Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ G-Tek™ GP™ Seamless Knit Nylon Gloves with Nitrile Coated MicroSurface Grip

Catalog Number p-4316967
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Seamless knit shell increases comfort and finger dexterity, while the MicroSurface coating is liquid impermeable to enhance grip in dry or oily conditions.

  • Seamless knit shell increases comfort, finger dexterity, and breathability
  • Nitrile MicroSurface coating is impermeable to provide an elevated grip in dry and oily conditions
  • Coating is infused with thousands of tiny suction cup pockets that create a vacuum effect to dispense fluids away on contact
  • Knit wrist helps prevent dirt and debris from entering the glove
  • Resistant to chemicals, water, and ultraviolet light
  • Washable
ANSI Abrasion Rating ANSI 1
Coated Area Palm, Fingers
Construction Seamless Knit
Coating Material Nitrile
Color Black, Green
Exterior Finish Fully Textured
Hand Specific
Material Nylon
Product Type Seamless Knit Gloves
Size X-Small
For Use With (Application) Oily Parts Assembly, Metal Handling, Fishing, and Machine Operation
Glove Cuff Style Knit
Series 34-400
Yarn Gauge 18
Length (English) 7.7 in.
Length (Metric) 19.5 cm
Show More Show Less
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit