Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ CleanTeam™ Seamless Knit Nylon Clean Environment Gloves with Polyurethane-Coated Fingers

Catalog Number p-3203449
Click to view available options
Size:
Medium
Large
X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Seamless nylon knit shell reduces hand fatigue, while the polyethylene coating provides a secure grip in dry or slightly wet/oily conditions.

  • Knit Wrist helps prevent dirt and debris from entering the glove
  • Low lint gloves are knit from nylon filament yarns
  • Nylon yarns absorb perspiration to leave hands cool and comfortable
  • Can be worn as a liner under disposable and other liquid-proof gloves
  • Color coded hems for easy size identification
Coated Area Fingertips
Construction Seamless Knit
Coating Material Polyurethane
Color White
Exterior Finish No Texture
Hand Specific
Material Nylon
Size Medium
For Use With (Application) Product Handling, Electronics Assembly, Photography Labs, and Computer Manufacturing
Glove Cuff Style Knit
Series 99-126
Show More Show Less

WARNING: Cancer - www.P65Warnings.ca.gov

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit