Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ CleanTeam™ Cleanroom Paper

For use in controlled environments

$624.00 - $743.00


Products 4
Catalog Number Mfr. No. Color Price Quantity & Availability  
Catalog Number Mfr. No. Color Price Quantity & Availability  
19-130-1954
View Documents
PIP™
10095501B
Blue
Case of 2500 for $743.00
Add to Cart
 
19-130-1955
View Documents
PIP™
10095501G
Green
Case of 2500 for $624.00
Add to Cart
 
19-130-1956
View Documents
PIP™
10095501P
Pink
Case of 2500 for $743.00
Add to Cart
 
19-130-1953
View Documents
PIP™
10095501W
White
Case of 2500 for $743.00
Add to Cart
 
Description

Description

  • Antistatic properties reduce attraction of airborne particles and enhance its performance in photocopiers and converting processes
  • Chemical and moisture resistant
  • Weight: 9.98 kg (22 lb.)
  • L × W: 21.6 × 27.9cm (8.5 × 11.0 in.)

SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit