Learn More
Zymo Research Corporation Pinpoint™ Solution (1 ml)
Supplier: Zymo Research Corporation D30011
Simply apply to Pinpoint Solution the desired section, allow to dry, and lift the desired section off the slide.