Learn More
Thermo Scientific™ Pierce™ Streptavidin Coated High Sensitivity Plates, Clear, 8-Well Strip
Maximize biotinylated protein capture efficiency and control background with microplates coated to detect low concentrations of biotinylated analytes.
Supplier: Thermo Scientific™ 15520
Alert:
Description
Thermo Scientific™ Pierce High Sensitivity Streptavidin Plates are biotin-binding microplates that are coated to maximize capture efficiency of biotinylated probes and control background in assays for low levels of target analyte.
These high sensitivity plates enable detection of low concentrations (ng/mL) of biotinylated antibodies and other proteins. Whether detecting a signal by fluorescent, colorimetric or chemiluminescent methods, these plates make it easy to develop an ELISA for any sample with a low-level target.Highlights:
Sensitive – detect down to 5ng/mL of biotinylated IgG
Specific – low background with high signal-to-noise ratios
Versatile – use either fluorescence, chemiluminescence or colorimetric detection
Robust – broad dynamic range
Ready to use – plates are supplied pre-blocked to save time
Specifications
High Sensitivity, Affinity | |
Pierce Streptavidin Coated High Sensitivity Plates, Clear, 96-Well Strip | |
5 Plate | |
Clear | |
Polystyrene | |
Streptavidin |
Upon receipt store at 4°C. Product is shipped at ambient temperature. | |
Fluorescence | |
Pierce™ | |
Microplate | |
96 | |
Blocker™ BSA Buffer |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.