Learn More
Thermo Scientific™ Pierce™ Biotin-Binding Plate Sample Pack
Determine which variety of streptavidin-coated plate is best for a particular assay involving biotinylated molecules with this sample pack.
Supplier: Thermo Scientific™ 15115
Description
Thermo Scientific™ Pierce Biotin-Binding Plate Sample Pack is ideal for establishing which variety of streptavidin coated plate is best for particular assay involving biotinylated molecules.
This sample pack contains streptavidin and NeutrAvidin™ Protein coated on microplates that are preblocked with either SuperBlock™, Blocking Buffer or Blocker™ BSA. This variety allows researchers to determine which plate produces the best signal-to-noise ratio for a specific experimental system. Once optimal conditions are empirically determined, the plate that provides the best results can be purchased in bulk for the remainder of the study.
Highlights:
- Two protein types: streptavidin and Thermo Scientific NeutrAvidin™ Protein
- Two blocking agents: bovine serum albumin (BSA) and Thermo Scientific SuperBlock™ Buffer
- Two coating volumes: 100μL per well and 200μL per well
Specifications
High Capacity, Standard Capacity, Affinity, Multiple | |
Fluorescence | |
Pierce™ | |
Coated Strip Plate | |
Streptavidin, NeutrAvidin Protein |
Store plates at 4°C. Store opened, unused plates in a resealable bag with desiccant at 4°C. Shipped at ambient temperature. | |
Sample Pack | |
Clear | |
96 | |
SuperBlock™ Buffer/Blocker™ BSA Buffer |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.