Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Picric Acid, Saturated, LabChem™

Click to view available options
Quantity:
500 mL
1 L
88-89-1, 7732-18-5
C6H3N3O7
229.104
OXNIZHLAWKMVMX-UHFFFAOYSA-N
picric acid, trinitrophenol, carbazotic acid, picronitric acid, melinite, acide picrique, phenol trinitrate, picral, nitroxanthic acid, pikrinsaeure
6954
CHEBI:46149
2,4,6-trinitrophenol
C1=C(C=C(C(=C1[N+](=O)[O-])O)[N+](=O)[O-])[N+](=O)[O-]
88-89-1, 7732-18-5
C6H3N3O7
229.104
OXNIZHLAWKMVMX-UHFFFAOYSA-N
picric acid, trinitrophenol, carbazotic acid, picronitric acid, melinite, acide picrique, phenol trinitrate, picral, nitroxanthic acid, pikrinsaeure
6954
CHEBI:46149
2,4,6-trinitrophenol
C1=C(C=C(C(=C1[N+](=O)[O-])O)[N+](=O)[O-])[N+](=O)[O-]
Color Yellow
CAS Max % 98.7, 1.3
Linear Formula C6H3N3O7
Solubility Information Soluble in water
Formula Weight 229.1
Density 1g/mL
Grade Certified
Quantity 1 L
Packaging Poly Bottle
Physical Form Liquid
Specific Gravity 1g/mL
Chemical Name or Material Picric Acid, Saturated
Show More Show Less

Recommended Storage : Room Temperature

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit