Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Aqua Phoenix™ Physical Changes Kit
Students learn about the physical changes of matter.
Supplier: Aqua Phoenix™ 1168
Alert:
CHOKING HAZARD (1). Small parts. Not for children under 3 yrs.
Includes: Instructor manual, student data sheets, ice cube trays, food coloring, plastic cups, cornstarch, measuring cups, rolling pins, shape molds, microwave popcorn and Play-Doh™ (four colors)
CHOKING HAZARD (1). Not for under 3 yr.
Requires : Microwave and clock.
This item is not returnable. View return policy
Description
Students observe physical changes of matter — size, shape or state—and see that these changes do not produce new substances
- Includes four activities
- Investigate what happens to matter as it absorbs or looses heat
- Explore changing the sizes and shapes of substances
- Grade K-4
- Class size: 24 students
Specifications
Physical Changes Kit | |
K to 4 | |
Students learn to observe and understand physical changes in the size, shape, or state of matter of a substance, without its producing or becoming a new substance | |
Instructor manual, student data sheets, ice cube trays, food coloring, plastic cups, cornstarch, measuring cups, rolling pins, shape molds, microwave popcorn and play-doh™ (four colors) |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction