Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phosphomolybdic Acid Hydrate, Honeywell Fluka™
Click to view available options
Quantity:
25 g
100 g
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Density | 1.62g/mL at 25°C |
Quantity | 25 g |
Linear Formula | H3[P(Mo3O10)4] · xH2O |
UN Number | 3084 |
Formula Weight | 1825.25g/mol (anhydrous basis) |
Grade | ACS Reagent |
Chemical Name or Material | Phosphomolybdic Acid Hydrate |
Safety and Handling
P220-P280-P305 + P351 + P338-P310
H272-H314
EINECSNumber : 234-713-5