Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phosphate Standard Solution, 1000 ppm P, Spectrum™ Chemical
SDP

Catalog Number 18612321
missing translation for 'orderingAttributeHoverText'
Quantity:
500 mL
Packaging:
Poly Bottle
7778-77-0
H2KO4P
136.08
MFCD00011401 MFCD00147253
GNSKLFRGEWLPPA-UHFFFAOYSA-M
potassium dihydrogen phosphate
[K+].OP(O)([O-])=O
7778-77-0
H2KO4P
136.08
MFCD00011401 MFCD00147253
GNSKLFRGEWLPPA-UHFFFAOYSA-M
potassium dihydrogen phosphate
[K+].OP(O)([O-])=O
Quantity 500 mL
Concentration or Composition (by Analyte or Components) 1000 ± 10 ppm
Packaging Poly Bottle
Appearance Clear, Colorless

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.

missing translation for 'provideContentCorrection'

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

missing translation for 'productTitle'