Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phenol, Saturated (pH 4.3, Liq.), Fisher BioReagents
Click to view available options
Quantity:
100 mL
400 mL
Packaging:
Coated Amber Glass/Poison Pack
Description
Phenol saturated with Tris buffer is used for purifying RNA.
Specifications
Assay Percent Range | ≥99.0 |
Density | 1.05g/cm³ |
Percent Purity | ≥99.0% |
Identification | Pass Test |
Packaging | Coated Amber Glass/Poison Pack |
pH | 4.3 |
Quantity | 400 mL |
Physical Form | Liquid |
Chemical Name or Material | Phenol, Saturated |
Safety and Handling
Hazard Statement | DANGER! |
Precautionary Statement | Emergency Overview Combustible liquid. Causes burns by all exposure routes. May cause central nervous system effects. Possible risks of irreversible effects. Danger of serious damage to health by prolonged exposure. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Immediate medical attention is required. Move to fresh air. NFPA Health:4 Flammability:2 Instability:0 |
DOTInformation | DOT Class 6.1, : Poison |
Recommended Storage | 4°C |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction