Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenol/Chloroform/Isoamyl Alcohol (125:24:1 Mixture, pH 4.3, Liq.), Fisher BioReagents™

Catalog Number BP1754I400
Click to view available options
Quantity:
100 mL
400 mL
Packaging:
Coated Amber Glass/Poison Pack
123-51-3
C5H12O
88.15
PHTQWCKDNZKARW-UHFFFAOYSA-N
3-methyl-1-butanol, isoamyl alcohol, isopentyl alcohol, isopentanol, 3-methylbutanol, 1-butanol, 3-methyl, isoamylol, isobutylcarbinol, iso-amylalkohol, isobutyl carbinol
31260
CHEBI:15837
3-methylbutan-1-ol
CC(C)CCO
123-51-3
C5H12O
88.15
PHTQWCKDNZKARW-UHFFFAOYSA-N
3-methyl-1-butanol, isoamyl alcohol, isopentyl alcohol, isopentanol, 3-methylbutanol, 1-butanol, 3-methyl, isoamylol, isobutylcarbinol, iso-amylalkohol, isobutyl carbinol
31260
CHEBI:15837
3-methylbutan-1-ol
CC(C)CCO

Phenol/Chloroform/Isoamyl Alcohol (125:24:1 Mixture, pH 4.3, Liq.)

This mixture is used to purify total RNA from mixtures containing RNA, DNA, and protein.
TRUSTED_SUSTAINABILITY
Merck Index 15, 7354
Percent Purity ≥99%
Identification Pass Test
Packaging Coated Amber Glass/Poison Pack
pH 4.3
Quantity 400 mL
Physical Form Liquid
Chemical Name or Material Phenol/Chloroform/Isoamyl Alcohol, 125:24:1 Mixture
DOTInformation DOT Class 6.1, : Poison
Recommended Storage 4°C
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit