Learn More
FLEXBIOSYS INC PHARMED MALE-FEMALE EXTENSION
Supplier: FLEXBIOSYS INC FXK00010444
FLEXREADY™ Pharmed Male-Female Extension,1/8" ID x 1/4" OD x 36", PharMed BPT, Single Pouch,Gamma Irradiated at 25 - 40 kGy., Rev A, Ea.