Learn More
Paul N Gardner Company PHARMACEUTICAL VOLUMETER 10 ME
Supplier: Paul N Gardner Company WG0603
Phamaceutical Volumeter with 10 mesh funnel , Used to determine apparent density of microcrystalline cellulose.The upper funnel contains a 10 mesh wire support, and Uses a 25cc brass density cup