Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Estes Industries, LLC Phantom
SDPSCI_ED

What better way to present the inner workings of a rocket and to help explain how rockets work than the Phantom Display Model Rocket Kit.

Supplier:  Estes Industries, LLC 001207

Catalog No. S68264


Add to Cart

Description

Description

  • Made with clear plastic parts
  • Only requires 15 minutes and glue to assemble
  • Phantom will become your go-to rocket for classroom rocket experiments, demonstrations and displays.
  • For display only. Not a flying model.
Specifications

Specifications

Siliver, Yellow, Blue, Pink
16.1 in.
12.1 x 10.8 x 12.1 in.
Beginner
30.7 cm
12.1 in.
Plastic
36.9 g
Model Rocket
0.98 in.
Rocket
12.1 in.
Plastic body parts, stickers, Included cutaway engine only. Non-flying model
30.7 cm
1.3 oz.
10.8 in.
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit