Learn More
Cayman Chemical Phalloidin, 17466-45-4, 1 mg
Supplier: Cayman Chemical 180391
A natural mycotoxin that binds filamentous actin (F-actin) and stabilizes actin fibers; commonly used to stain F-actin in cells when fluorescently labeled