Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ PH111 pH/mV Bench Meter with Stand
SCI_ED

Orion Lab Star PH111 pH bench meters measure pH or ORP with mV and temperature, feature a large backlit display for easy viewability, and offer small footprint to save bench space. Ideal for simplified, budget-friendly lab testing and data recording.

Supplier:  Thermo Scientific™ LSTAR1110/EMD

Catalog No. S81896


Add to Cart

Description

Description

Orion Lab Star PH111 pH Meter Features

  • Budget-friendly pH meter for simplified pH and ORP testing
  • Display shows pH, mV and temperature simultaneously for accurate data recording
  • Measure -2 to 18 pH, -2000 to +2000 mV and -5 to 105 °C or 23 to 221 °F to ensure comprehensive measurement capabilities
  • Perform one to five point pH calibrations, one point ORP calibrations and one point temperature adjustment calibrations for proficient SOP compliance
  • Large 5 inch backlit display makes viewing and recording measurements easy
  • Small meter footprint saves valuable bench space in your lab
  • Onscreen stabilizing and ready indicators help identify when readings are complete
  • pH electrode condition icon helps ensure reliable electrode performance
  • Date and time data stamped data and calibration logs support GLP compliance
  • View active pH, ORP and temperature calibration logs with date and time stamp to ensure accurate, reproducible measurements
  • Use 500-point data log with date and time stamp to record key measurements
  • Output data and calibration logs to a printer or computer for long-term data storage
  • Interchangeable wall plugs for US/Japan, Euro, UK/Singapore, Australia/New Zealand and China outlets

Meter and Meter Kits

  • LSTAR1110 is the pH meter and stand option, which allows you to purchase your pH electrode separately
  • LSTAR1115 is the standard pH meter kit option, for common pH samples such as drinking water, tap water, buffers (non-TRIS) and general purpose student use
  • LSTAR116 is the easy-to-clean bio pH meter kit option, for pH samples containing TRIS buffers, proteins, sulfides, halides or other components that can interfere with silver-chloride; included refillable pH electrode is easy-to-clean and restore
  • LSTAR117 is the low-maintenance bio pH meter kit option, for pH samples containing TRIS buffers, proteins, sulfides, halides or other components that can interfere with silver-chloride; included gel-filled pH electrode is low-maintenance and easy to operate
  • LSTAR1118 is the difficult sample pH meter kit option, for pH samples that are thick, viscous or semi-solid including foods, beverages, wastewaters, emulsions, soils, slurries, suspended solids or sludges
Specifications

Specifications

pH/mV Bench Meter with Stand
±0.2% or ±0.05% (Whichever is Greater)
Transfer Single or All Data Points to Printer or Computer
Meter with Electrode Stand, PC Cable, Power Adapter
pH with mV and Temperature, ORP with mV and Temperature
-2 to 18
1 to 5 Points
0.1 mV
-5°C to +105°C
0.1°C (0.1°F)
1 Point Offset Calibration
Yes
500 Points with Date and Time Stamp
Calibration Reminder Alarm
CE, TUV 3-1, FCC Class A
198.3 x 155.2 x 61.7 mm (7.81 x 6.11 x 2.43 in.)
1.54 lb.
Universal
50/60 Hz
±0.3°C (±0.5°F)
±0.005, ±1 LSD
Computer or Printer (STARA-106)
Allows for Purchasing Electrode Separately
pH, ORP, Temperature
0.001, 0.01, 0.1
±2000 mV with ±250 mV Offset
1 (ORP)
23°F to 221°F
Manual, Automatic
LCD, Backlit
Non-volatile
Automatic with Auto-read and Timed Read Modes, Manual with Continuous Read Mode
BNC for pH or ORP, 8-pin MiniDIN for ATC
IP54
700 g
3 Years (Meter)
100/240 V
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit