Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisherbrand™ pH Paper Rolls
General purpose pH papers in efficient 15' rolls
Description
- Convenient dispenser allows you to select the right amount of pH paper for each application
- Easy-to-usedash (em)Dip paper in solution to be tested and match to color chart immediately
- Highly accurate and well suited for most applications
- Each SKU is packed in box of five clear plastic dispensers
- Cat. No. 13-640-507 is a pH paper testing set that includes shrink wrapped pack of seven dispensers (one of each SKU)
- Store at room temperature. Keep out of direct sunlight.
Order Info
15 ft. Roll provides about 100 tests.
Specifications
0 to 14 | |
Includes 1 each of 13640500, 13640501, 13640502, 13640503, 13640504, 13640505, 13640506, | |
Room Temperature |
Convenient dispenser allows to select the right amount of pH paper for each application, highly accurate and well suited for most applications | |
7/Pk. | |
pH Test Paper Set |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction