Learn More
CTL Scientific Supply Corp PH-FIX STRIPS 6-10,CS/10BX/100
Supplier: CTL Scientific Supply Corp 9212210
PH-FIX 6.0-10.0 TEST PAPER are non-bleeding indicator strips. Gradations of scale: 6.0 - 6.4 - 6.7 - 7.0 - 7.3 - 7.6 - 7.9 - 8.2 - 8.4 - 8.6 - 8.8 - 9.1 - 9.5 - 10 (case of 10 boxes of 100 strips each)