Learn More
CTL Scientific Supply Corp PH-FIX STRIPS 0-14,CS/10BX/100
SDP
Supplier:  CTL Scientific Supply Corp 9211010
Encompass_Preferred
PH-FIX 0-14 TEST PAPER are non-bleeding indicator strips. Gradation of scale: 0 - 1 - 2 - 3 - 4 - 5 - 6 - 7 - 8 - 9 - 10 - 11 - 12 - 13 - 14 (case of 10 boxes of 100 strips each)
Catalog No. 50-117-3340