Learn More
CTL Scientific Supply Corp PH-FIX 3.1-8.3,100 TEST STRIPS
Supplier: CTL Scientific Supply Corp 92135
PH-FIX 3.1-8.3 are non-bleeding indicator strips. Gradation of scale: 3.1 - 3.5 - 3.9 - 4.3 -4.7 - 5.1 - 5.5 - 5.9 - 6.3 - 6.7 - 7.1 - 7.5 - 7.9- 8.3, box of 100 strips