Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
pH Buffer Standard Solutions pH 10.012, Hach Lange™
Click to view available options
Quantity:
500 mL
Description
- pH Standard, IUPAC, pH10.012 at 25 °C, Certified, 500 mL (Radiometer Analytical)
- 10.012 ± 0.010 pH at 25°C concentration
- Traceable to IUPAC pH scale and NIST
- Shelf life of 48 months from production date
- Each bottle (500 mL) is supplied with a COFRAC Verification Certificate and Certificate of Conformity and Traceability drawn up according to ISO31
- Total Control of the traceability chain and calculated uncertainties
- Direct traceability to primary standards and the reference method
- The competence and integrity of a laboratory internationally recognized to be accredited for the measurement of pH and conductivity reference materials
Specifications
Melting Point | 0°C |
Color | Colorless |
pH | 10.012 |
Physical Form | Liquid |
Quantity | 500 mL |
Odor | Odorless |
Traceability to NIST | Yes |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction