Learn More
Miele, Inc. PG 8504 208V Undercounter Labwasher
Supplier:  Miele, Inc. 10250010
Encompass_Preferred
Laboratory glassware washer with DI water supply.Variable-speed heater pumpOptimum cleaning performanceLow consumption valuesOptimum wash pressureThis laboratory glassware washer reengineered its undercounter glassware washer line, taking glassware reprocessing to a higher level with the new cutting-edge Flex Series Washers. Featuring a completely redesigned wash chamber as well as a powerful variable-speed pump, this new undercounter glassware washer provide more capacity, greater load flexibility and more efficient application specific programs.The PG 8504 laboratory glassware washer is a 23.6" wide unit with 3 wash programs, stainless steel interior and exterior and an indoor powder detergent, as well as neutralizer dispenser. External DOS module for liquid detergent dispensing can be added.
Catalog No. NC1609532