Learn More
Promega pFC14K HaloTag™ CMV Flexi™ Vector
Vectors designed for expression of C-terminal-tagged HaloTag™ fusion proteins in mammalian cells. Once expressed, use the HaloTag™ fusion protein for cell imaging of protein localization or trafficking in conjunction with the fluorescent HaloTag™ Ligands.
Supplier: Promega G9661
Description
- Appends HaloTag(R) to C-Terminus of Protein Fusion Partner
- Enables constitutive protein expression in mammalian cells
- Includes CMV immediate-early enhancer/promoter
- Suited to stable or transient gene expression
Specifications
-30°C to -10°C | |
20 μg |