Learn More
Dynalon™ Petri Dishes
For culturing various organisms. Dynalon™ Petri Dishes are of two types. No vent limits oxygen uptake and with 4 vent allows for oxygen uptake.
Supplier: Dynalon™ 4020240000
Description
- Made of transparent polystyrene which are optically clear
- Stackable and thus reduces storage space
- For single use
- Consistent flatness allows for even media distribution
- Certificate of Sterilization upon request
- 402024-0000 has no vent and limits oxygen uptake. Gamma irradiated
Specifications
Stackable | |
Disposable | |
Round | |
No |
Clear | |
Polystyrene | |
Sterile |