Learn More
CTL Scientific Supply Corp PEROXTESMO KO,100 PIECES
Supplier: CTL Scientific Supply Corp 90606
This test paper allows the quick and easy detection of peroxidase in food. It is used in the food processing industry to evaluate the quality of canned food. Color reaction: white, blue-green