Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3B Scientific™ Periodic Table of the Elements, With Electron Configurations
3B Scientific™ Periodic Table of the Elements, with Electron Configurations is displayed on strong laminated material with rods and a hanging cord
Supplier: 3B Scientific™ U197001
Specifications
Poster | |
54.3 in. | |
Plastic | |
English, German |
78.6 in. | |
1 | |
White, Orange, Green, Blue | |
Chemistry |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction