Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HexArmor™ Rig Lizard Arctic™ 2023 Cold Weather Work Gloves
Click to view available options
Small
Description
- H2X™ waterproof and C40 Thinsulate™ interior liners keep hands warm and dry
- Durable TP-X™ palm provides the enhanced durability and grip capabilities
- Back-of-hand IR-X™ Impact Exoskeleton™ with a high-flex design keeps hands safe from smash & impact hazards
- Gloves provide workers with ANSI/ISEA 138 Level 1 impact protection
- Reinforced index finger and thumb crotch increase longevity
- SlipFit™ cuff increases comfort and convenience
- Hi-vis color scheme is ideal for low-light applications
- Machine washable
Specifications
Back Material | IR-X™ Impact Exoskeleton™ |
Certifications/Compliance | ANSI/ISEA 138 Level 1 Impact Protection |
Cuff Material | SlipFit™ |
Coating Material | Uncoated |
Color | Red, Hi-vis Yellow |
Cut Resistance Rating | ANSI A3 |
Hand | Specific |
Size | Small |
Liner Material | H2X™, C40 Thinsulate™ |
Palm Type | TP-X™ |
Show More |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction