Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Performance Check Solution, SPEX CertiPrep™

Click to view available options
Quantity:
1mL
Packaging:
2mL amber prescored glass ampule
50-29-3
C14H9Cl5
354.476
YVGGHNCTFXOJCH-UHFFFAOYSA-N
3036
CHEBI:16130
1-chloro-4-[2,2,2-trichloro-1-(4-chlorophenyl)ethyl]benzene
C1=CC(=CC=C1C(C2=CC=C(C=C2)Cl)C(Cl)(Cl)Cl)Cl
50-29-3
C14H9Cl5
354.476
YVGGHNCTFXOJCH-UHFFFAOYSA-N
3036
CHEBI:16130
1-chloro-4-[2,2,2-trichloro-1-(4-chlorophenyl)ethyl]benzene
C1=CC(=CC=C1C(C2=CC=C(C=C2)Cl)C(Cl)(Cl)Cl)Cl

Includes

Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
Aldicarb sulfoxide1646-87-3100μg/mL
Methiocarb2032-65-720μg/mL
4-Bromo-3,5-dimethylphenyl-N-methylcarbamate672-99-110μg/mL
3-Hydroxycarbofuran16655-82-62μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Packaging 2mL amber prescored glass ampule
Methods For US EPA Method 531.1
Solvent or Matrix Methanol
Quantity 1mL
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
Concentration or Composition Notes Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity.
Chemical Name or Material Performance Check Solution, USEPA Method 531.1

DOTInformation : Nonhazardous

Recommended Storage : Ambient

Product intended for laboratory use only