Learn More
Sigma Aldrich Fine Chemicals Biosciences Percoll, MFCD00131830, 100mL
Supplier: Sigma Aldrich Fine Chemicals Biosciences P4937100ML
pH 8.5-9.5 (25 °C), suitable for cell culture, Percoll consists of colloidal silica particles of 15-30 nm diameter (23% w/w in water) which have been coated with polyvinylpyrrolidone (PVP).