Learn More
Chemglass Life Sciences 500mL Peptide Synthesis Vessel, Solid Phase, Coarse Frit, GL 32 Thread
Supplier: Chemglass Life Sciences CG186015
500mL Peptide Synthesis Vessel, Solid Phase, Coarse Frit, GL 32 Thread. Solid phase peptide synthesis vessel having a medium or coarse porosity fritted glass resin support. Top of the vessel has a GL thread and is supplied complete with a PTFE lined PBT screw cap. Lower PTFE stopcock has a 2mm bore with the arm being 8mm O.D. Vessels are available in sizes other than those listed. Please call for details.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.