Learn More
Chemglass Life Sciences 25mL Peptide Synthesis Vessel, Solid Phase, Coarse Frit, GL 25 Thread
SDP
Supplier:  Chemglass Life Sciences CG186011
Encompass_Preferred
25mL Peptide Synthesis Vessel, Solid Phase, Coarse Frit, GL 25 Thread. Solid phase peptide synthesis vessel having a medium or coarse porosity fritted glass resin support. Top of the vessel has a GL thread and is supplied complete with a PTFE lined PBT screw cap. Lower PTFE stopcock has a 2mm bore with the arm being 8mm O.D. Vessels are available in sizes other than those listed. Please call for details.
Catalog No. CG186011