Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pentane, HPLC Grade, Spectrum™ Chemical

Click to view available options
Quantity:
1 L
4 L
Packaging:
Amber Glass Bottle
Specifications
CAS Max % | 1 |
Grade | High Purity |
Quantity | 4 L |
Packaging | Amber Glass Bottle |
Residue after Evaporation | 5 ppm |
Water | 0.0002 |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.