Learn More
CTL Scientific Supply Corp PEHANON PH 6-8.1,PK/600 STRIPS
Supplier: CTL Scientific Supply Corp 904173
Pehanon pH 6.0-8.1 indicator papers, pack of 2 boxes of 200 strips. The indicator and color scale are combined on a single strip. Gradation of scale: 6.0 - 6.3 - 6.6 - 6.9 - 7.2 - 7.5 - 7.8 - 8.1