Learn More
CTL Scientific Supply Corp PEH. PH 7.2-8.8 ,PK/600 STRIPS
Supplier: CTL Scientific Supply Corp 904193
Pehanon pH 7.2-8.8 indicator papers, pack of 2 boxes of 200 strips. The indicator and color scale are combined on a single strip. Gradation of scale: 7.2 - 7.4 - 7.6 - 7.8 - 8.0 - 8.2 - 8.5 - 8.8