Learn More
ELLARD INSTRUMENTATION LTD Pediatric Stadiometer
Supplier: ELLARD INSTRUMENTATION LTD PED LB 35107X
Useful in a variety of clinical settings. Designed for use by doctors, nurses and nutritionists in pediatric practice. Measures recumbent length of all children up to 2 years of age and many to 3 years.Measures from 35-107 cm and 14-42 inches. Stationary, 90º angle head piece. Removable, sliding, 90º angle foot piece with magnifier. Easy to read measuring tape in English and metric Calibration in 1/8 inch and 1 mm increments. Meets CHDP specification for state of California Made of clear acrylic reinforced with 1/4" white ABS. Weight: 6 pounds. Built-in handle for easy transport Overall length: 49 inches
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.