Learn More
Air Science Usa Air Science Purair™ PCR Workstation, 48" / 1200mm Nominal Width, 115V 60Hz
Supplier: Air Science Usa PCR48
Purair PCR Workstation with vertical laminar flow and UV (PCR-48), 48" / 1200mm nominal width. Polymerase chain reaction (PCR) is a simple and inexpensive technique to make multiple copies of a targeted nucleotide sequence from a DNA sample and to amplify sequences from small samples. This technique is widely used in genetics laboratories that work with DNA and RNA. Because PCR amplification is extremely sensitive to contamination, prevention of contamination requires good laboratory practices to minimize external or cross-contamination during reagent preparation, sample preparation and sample amplification. The Purair PCR laminar flow cabinet employs the Air Science Multiplex HEPA filtration technology to create a safe, energy-efficient contaminant-free environment. This PCR workstation is ideally suited for use with non-hazardous contaminants and when flexible access to the instrumentation in the cabinet is required
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.