Learn More
BrandTech™ BRAND™ PCR 12-Strip Tubes
Manufactured from polypropylene without lubricants or chemical additives that can contaminate reagents and samples.PCR 12-Strip Tubes are autoclavable and RNase-, DNase-, and DNA-free. Thin, uniform walls to ensure consistent, precise thermal transfer for excellent results. Available with or without caps and can be ordered separately.
Supplier: BrandTech™ BRAND™ 781282
Description
Manufactured from high-quality virgin plastics for excellent, reproducible results in most PCR applications, and are manufactured without lubricants or chemical additives that can contaminate reagents and samples.- 12-strip tube strips are available with or without caps. 12-strip caps can be ordered separately.
- Thin, uniform walls to ensure consistent, precise thermal transfer for excellent results.
- Autoclavable
- RNase-, DNase-, and DNA-free.
- Compatible with most 24-, 32-, 96-, and 384-well thermal blocks.
- Available in convenient tube/cap sets.
Specifications
Certified RNase/DNase-free | |
Without Lid | |
Yellow | |
0.2 mL | |
125/Pk. |
Strip Plate | |
Autoclavable | |
Polypropylene | |
Yes | |
Yes |