Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Paulson Shield Labels
Provide positive identification at a distance
Specifications
Color | Black on White |
---|---|
Material | Mylar |
Diameter (Metric) | 60.96 cm |
For Use With (Equipment) | Body and capture shield |
Weight (English) | 4 oz. |
Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
19-500-351
|
Paulson
BSLABC |
Body Shield Identification Label: Corrections | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
19-500-353
|
Paulson
BSLABP |
Body Shield Identification Label: Police | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
19-500-352
|
Paulson
BSLABM |
Body Shield Identification Label: Military Police | Item Discontinued This item has been discontinued by Fisher Scientific. Please Sign In to view product availability in your area. | N/A | |||||
19-500-355
|
Paulson
BSLABS |
Body Shield Identification Label: Sheriff | Item Discontinued This item has been discontinued by Fisher Scientific. Please Sign In to view product availability in your area. | N/A | |||||
19-500-354
|
Paulson
BSLABPA |
Body Shield Identification Label: Policia | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
Description
- Replacement labels for body and capture shields
- Mylar™ self-adhesive labels attach to inside surface of shield
Specifications
Black on White | |
60.96 cm | |
4 oz. |
Mylar | |
Body and capture shield |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title