Learn More
Paulson Industrial Chemical Goggles
Click to view available options
Lexan lens; Black with yellow bezel; 5.75 oz
Specifications
Product Type | Industrial Chemical Goggles |
Frame Color | Black with yellow bezel |
Ventilation Details | Bottom vents |
Lens Material | Lexan plastic |
Frame Material | Silicone |