Learn More
Paulson A-TAC™ “FRAG” Goggles (TF Series)
Click to view available options
Frag
Description
Includes
Include mounting clips for PASGT (Personnel Armor System, Ground Troops) helmets.
A-TAC Frag
- Provides fragmentation protection
- Triple lenses with hard-coated outer lens, center fragmentation lens and anti-fog inner spall lens
- V-50 rated
- Weighs 6.75 oz.
Order Info
Noseguard option available for A-TAC Frag. Contact your Fisher Safety Customer Service Representative for ordering information.
Compliance
Meet ANSI Z87.1 standards.
Specifications
Certifications/Compliance | ANSI Z87.1 |
Lens Tint | Clear |
Product Type | Tactical Goggle |
Includes | Black Strap |
Frame Material | Silicone |