Learn More
Paulson A-TAC™ Firefighter Goggles: Structural
Click to view available options
Product Type:
Firefighter Goggles
Firefighter goggle w/quick relaese buckle
Description
- Polycarbonate, hard-coated/anti-fog lens
- Fit comfortably over prescription, sun, and safety glasses
- Indirect ventilation ports
- Can be placed on the helmet or worn on the head
Model 510EB
- Silicone frame
- Optically correct lens
- Quick-release buckle
- Apec lens
- Black frame
With noseguard
Compliance
Meets ANSI Z87.1 standard for impact, penetration and chemical splash exposure
Specifications
Certifications/Compliance | ANSI Z87.1 |
Description | Apec lens |
Lens Tint | Clear |
Product Type | Firefighter goggle w/quick relaese buckle |
For Use With (Application) | Wildland, structural or intense heat fire fighter |
Frame Color | Black |